Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

445820

Sigma-Aldrich

Meriolin 2

Meriolin 2 primarily used in Inhibition.

Synonym(s):

Meriolin 2, 3-((2-Amino)pyrimidin-4-yl)-4-hydroxy-1H-pyrrolo[2,3-b]pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9N5O
Molecular Weight:
227.22
UNSPSC Code:
12352200

Quality Level

Assay

≥93% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable, ATP-binding site-targeting azaindolopyrimidine compound that inhibits against Cdk2/A, Cdk9/T, DYRK1A, Cdk5/p25, CK1δ/ε, Cdk1/B, and GSK-3α/β kinase activity (IC50 = 18, 18, 35, 50, 50, 57, and 500 nM, respectively). Shown to induce DEVDase activity in SH-SY5Y neuroblastoma cultures (by 5.46-fold in 24 h at 1 µM) and exhibit antiproliferative activity toward SH-SY5Y, multiple myeloma KMS-11, human embryonic kidney HEK293, colorectal adenocarcinoma LS 174T, and glioma GBM cells in vitro (IC50 = 0.41, 0.6, 0.96, 1.06, and 1.21 µM, respectively).

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Echalier, A., et al. 2008. J. Med. Chem.51, 737.
Bettayeb, K., et al. 2007. Cancer Res.67, 8325.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service