Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

454585

Sigma-Aldrich

NMDA Receptor NR2BCT(1292-1304)-TAT

A 13-mer NMDA receptor 2B (NR2B) subunit carboxy terminal (1292-1304) derived peptide that specifically binds DAPK1 in a reversible manner and antagonizes the activated DAPK1-NR2B interaction and the potentiation of NMDAR function

Synonym(s):

NMDA Receptor NR2BCT(1292-1304)-TAT, H-KKNRNKLRRQHSY-Ahx-YGRKKRRQRRR-NH₂, Tat-NR2B CT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C143H254N64O32
Molecular Weight:
3381.96
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A 13-mer NMDA receptor 2B (NR2B) subunit carboxy terminal (1292-1304) derived peptide that specifically binds DAPK1 (death-associated protein kinase 1) in a reversible manner and antagonizes the activated DAPK1-NR2B interaction and the potentiation of NMDAR function with no effect on either the catalytic activity of DAPK1 or the NMDA receptor physiology. Blocks Ca2+ influx through extrasynaptic NMDA receptors (at 50 µM) and the blood-brain-barrier permeable version, namely NR2BCT(1292-1304) attached to TAT(47-57) via a spacer (6-aminohexanoic acid) protects cortical neurons from NMDA receptor-mediated insults and reduces brain infarction and improve neurological functions in mice (10 mg/kg, i.v.).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

H₂N-Lys-Lys-Asn-Arg-Asn-Lys-Leu-Arg-Arg-Gln-His-Ser-Tyr-Ahx-Tyr-Gly-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg-CONH₂

Physical form

Supplied as a trifluoroacetate salt

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Tu, W., et al. Cell140, 222.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service