475843
Minocycline, Hydrochloride - CAS 13614-98-7 - Calbiochem
Semi-synthetic tetracycline derivative effective against tetracycline-resistant staphylococci.
Synonym(s):
Minocycline, Hydrochloride - CAS 13614-98-7 - Calbiochem, 7-Dimethylamino-6-demethyl-6-deoxytetracycline, HCl
About This Item
Recommended Products
Quality Level
Assay
≥98% (TLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
dark yellow
solubility
water: 50 mg/mL
shipped in
ambient
storage temp.
2-8°C
InChI
1S/C23H27N3O7.ClH/c1-25(2)12-5-6-13(27)15-10(12)7-9-8-11-17(26(3)4)19(29)16(22(24)32)21(31)23(11,33)20(30)14(9)18(15)28;/h5-6,9,11,17,27-28,32-33H,7-8,24H2,1-4H3;1H/b22-16-;/t9-,11-,17-,23-;/m0./s1
InChI key
KDLQIOPKJDNQIM-WUURTAMISA-N
General description
Semi-synthetic tetracycline derivative effective against tetracycline-resistant staphylococci. A matrix metalloproteinase (MMP) inhibitor shown to inhibit tumor-induced angiogenesis. Inhibits stromelysin (MMP-3; IC50 = 290 M) but does not inhibit other MMPs such as fibroblast collagenase or gelatinase. Potently inhibits PARP-1 (Ki = 13.4 nM) in a NAD+ - competitive manner. Exhibits anti-inflammatory effects and suppresses T cell proliferation.
Biochem/physiol Actions
Metalloproteinase (MMP)
Warning
Other Notes
Gilbertson-Breading, S., et al. 1995. Cancer Chemother. Pharmacol. 36, 418.
Kloppenburg, M., et al. 1995. Clin. Exp. Immunol. 102, 635.
Preus, H.R., et al. 1995. J. Clin. Periodontol. 22, 380.
Weingart, J.D., et al. 1995. J. Neurosurg. 82, 635.
Tamargo, R.J., et al. 1991. Cancer Res. 51, 672.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service