Skip to Content
MilliporeSigma

495621

Sigma-Aldrich

Olomoucine II

A cell-permeable 2,6,9-trisubstituted purine analog that acts as a potent, reversible, and ATP-competitive inhibitor of Cdk1/B.

Synonym(s):

Olomoucine II, 6-(2-Hydroxybenzylamino)-2-((1R)-(hydroxymethyl)propyl)amino)-9-isopropylpurine

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

About This Item

Empirical Formula (Hill Notation):
C19H26N6O2
CAS Number:
Molecular Weight:
370.45
MDL number:
UNSPSC Code:
12352200

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

colorless

solubility

DMSO: 1 mg/mL
ethanol: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C19H26N6O2/c1-4-14(10-26)22-19-23-17(20-9-13-7-5-6-8-15(13)27)16-18(24-19)25(11-21-16)12(2)3/h5-8,11-12,14,26-27H,4,9-10H2,1-3H3,(H2,20,22,23,24)

InChI key

NDUVSANREQEDRE-UHFFFAOYSA-N

General description

A cell-permeable 2,6,9-trisubstituted purine analog that acts as a potent, reversible, and ATP-competitive inhibitor of Cdk1/B with an IC50 of 20 nM and displays increased potency over purvalanol A (Cat. No. 540500; IC50 = 50 nM), Roscovitine (Cat. No. 557360; IC50 = 450 nM), Bohemine (Cat. No. 203600; IC50 = 1.1 µM) and Olomoucine (Cat. No. 495620; IC50 = 7.0 µM). Further, exerts greater in vitro cytotoxicity in various cancer cell lines (IC50 = 3.0 µM, 5.3 µM, 6.3 µM, 6.3 µM and 11.1 µM for CEM, MCF7, HOS, G631 and K562, respectively).
A cell-permeable, potent, reversible, and ATP-competitive inhibitor of Cdk1/cyclin B (IC50 = 20 nM) that displays increased potency over purvalanol A (Cat. No. 540500; IC50 = 50 nM), Roscovitine (Cat. No. 557360; IC50 = 450 nM), Bohemine (Cat. No. 203600; IC50 = 1.1 µM), and Olomoucine (Cat. No. 495620; IC50 = 7.0 µM). Also exhibits greater in vitro cytotoxicity in various cancer cell lines (IC50 = 3.0 µM, 5.3 µM, 6.3 µM, 6.3 µM, and 11.1 µM for CEM, MCF7, HOS, G631, and K562, respectively).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Cdk1/B
Product competes with ATP.
Reversible: yes
Target IC50: 20 nM against Cdk1/B

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Krystof, V., et al. 2002. Bioorg. Med. Chem. Lett.12, 3283.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service