Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

497985

Sigma-Aldrich

Ornithine Decarboxylase Inhibitor, POB - Calbiochem

Synonym(s):

N-(4ʹ-Pyridoxyl)-Ornithine(BOC)-OMe, ODC Inhibitor, POB, Ornithine Decarboxylase Inhibitor, POB - Calbiochem

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$162.00

$162.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$162.00

About This Item

Linear Formula:
C19H31N3O6
UNSPSC Code:
12352200
NACRES:
NA.28

$162.00


Please contact Customer Service for Availability

Request a Bulk Order

form

(White to off-white solid)

Quality Level

storage condition

OK to freeze

solubility

soluble (H2O (1 mg/ml))

storage temp.

2-8°C

General description

A cell-permeable pyridoxyl-ornithine compound that acts as an effective analog of the transition state coenzyme-substrate adduct during orthithine decarboxylase- (ODC) catalyzed conversion of L-orthithine to Pultrescine. POB is typically 5- to 10-fold more potent than DFMO (Cat. No. 288500) in inhibiting intracellular ODC activity and tumor cell growth in cultures, while exhibiting little effect against the growth of nontumorigenic human aortic smooth muscle cells.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service