Skip to Content
MilliporeSigma

506156

Sigma-Aldrich

p38 MAP Kinase Inhibitor V

The p38 MAP Kinase Inhibitor V, also referenced under CAS 271576-77-3, controls the biological activity of p38 MAP Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

p38 MAP Kinase Inhibitor V, 4-(3-(4-Chlorophenyl)-5-(1-methylpiperidin-4-yl)-1H-pyrazol-4-yl)pyrimidine, SC-409

Sign Into View Organizational & Contract Pricing

Select a Size


About This Item

Empirical Formula (Hill Notation):
C19H20ClN5
CAS Number:
Molecular Weight:
353.85
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light yellow

solubility

DMSO: 10 mg/mL
ethanol: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C19H20ClN5/c1-25-10-7-14(8-11-25)19-17(16-6-9-21-12-22-16)18(23-24-19)13-2-4-15(20)5-3-13/h2-6,9,12,14H,7-8,10-11H2,1H3,(H,23,24)

InChI key

WYWOBCJMDNZHAV-UHFFFAOYSA-N

General description

A cell-permeable trisubstituted pyrazole compound that acts as a highly potent ATP-competitive inhibitor of CK1δ and p38α (IC50 = 20 and 40 nM, respectively). Shown to affect p38β, PKA, JNK2, and JNK3 activity (IC50 = 2.3, 10.5, 19.7, and 69.7 µM, respectively) only at much higher concentations and exhibit little effect against a panel of 44 other commonly studied kinases, including p38δ/γ, even at a conentraion of 10 µM or higher. An excellent tool for studying p38-mediated responses both in cells in vitro and in animals in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
p38 MAP Kinase
Product competes with ATP.
Reversible: no
Target IC50: 20 and 40 nM, against CK1δ and p38α, respectively

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Mbalaviele, G., et al. 2006. J. Pharm. Exp. Ther.317, 1044.
Liu, Y.H., et al. 2005. J. Card. Fail.11, 74.
Yu, J.J., et al. 2003. J. Immunol.171, 6112.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service