Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

521270

Sigma-Aldrich

PDK1, His•Tag®, Human, Recombinant, S. frugiperda

Synonym(s):

3-Phosphoinositide-Dependent Protein Kinase-1

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
2.7.10
UNSPSC Code:
12352202

Assay

≥90% (SDS-PAGE)

Quality Level

form

liquid

specific activity

≥75 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Full length, recombinant, human PDK1 fused to a His•Tag sequence and expressed in S. frugiperda insect cells. PDK1 is a serine/threonine protein kinase that is localized to the plasma membrane. PDK1 activates conventional protein kinase C (PKC) as well as PKCζ. PDK1 also phosphorylates Akt and plays a role in insulin signaling.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will transfer 1 nmol of phosphate to PDK1 substrate per min at 30°C, pH 7.5.

Physical form

In 100 mM NaCl, 50 mM Tris-HCl, 5 mM DTT, 500 µM EDTA, 500 µM EGTA, 20% glycerol, 0.05% TRITON X-100 Detergent, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Scott, M.T., et al. 2002. EMBO J.21, 6771.
Biondi, R.M., et al. 2000. EMBO J.19, 979.
Alessi, D.R., et al. 1997. Curr. Biol.7, 776.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service