Skip to Content
MilliporeSigma

526523

Sigma-Aldrich

PIM1/2 Kinase Inhibitor V

The PIM1/2 Kinase Inhibitor V, also referenced under CAS 327033-36-3, controls the biological activity of PIM1/2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PIM1/2 Kinase Inhibitor V, (Z)-5-(3-Trifluoromethylbenzylidene)thiazolidine-2,4-dione, SMI-4a, SMI-4a, (Z)-5-(3-Trifluoromethylbenzylidene)thiazolidine-2,4-dione

Sign Into View Organizational & Contract Pricing

Select a Size


About This Item

Empirical Formula (Hill Notation):
C11H6F3NO2S
CAS Number:
Molecular Weight:
273.23
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light beige

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C11H6F3NO2S/c12-11(13,14)7-3-1-2-6(4-7)5-8-9(16)15-10(17)18-8/h1-5H,(H,15,16,17)/b8-5+

InChI key

NGJLOFCOEOHFKQ-VMPITWQZSA-N

General description

A cell-permeable thiazolidinedione compound that acts as a potent, ATP-competitive inhibitor against Pim-1/2 kinases (IC50 = 24 nM and 100 nM against Pim-1 and Pim-2, respectively), while exhibiting little or no activity against a panel of 58 other kinases (≤20% inhibition at 5 µM). One hour inhibitor treatment (0.5 µM) of HEK-293T cultures at the end of a 4 hour 32PO4 incubation period results in nearly complete inhibition of the autophosphorylation of overexpressed Pim-1. Comparing to Inhibitor VI (Cat. No. 526524 ), Inhibitor V is less reactive towards DYRK1α (20% vs. 68% inhibition with respective inhibitor at 5 µM) and more potent in PC3 human prostate cancer cell killing (IC50 = 17 vs. 48 µM with respective inhibitor).

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Lin,Y., et al. 2009. Blood In press.
Xia, Z., et al. 2009. J. Med. Chem.52, 74.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service