Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

528112

Sigma-Aldrich

PI 3-Kγ/CKII Inhibitor

Synonym(s):

PI 3-Kγ/CKII Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.28

form

(Yellow solid)

Quality Level

storage condition

OK to freeze

solubility

soluble (DMSO (20 mg/ml))

storage temp.

2-8°C

General description

A cell-permeable furanyl-thiazolidinedione compound that acts as a potent, ATP-competitive inhibitor of PI 3-Kγ and CKII (IC50 = 20 nM). It inhibits other PI 3-K isotypes (IC50 = 0.94, 20, and 20 µM for α, β, δ, respectively) and MKK7β (IC50 >10 µM) only at much higher concentrations and exhibits little activity against 77 other commonly studied kinases even at concentrations as high as 10 µM. Shown to inhibit stimulated Akt phosphorylation in cells in vitro and offer blockage of thioglycollate-induced neutrophil recruitment in a murine peritonitis model in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service