Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

530007

Sigma-Aldrich

Prionogenesis inhibitor, DAPH-12

The Prionogenesis inhibitor, DAPH-12, also referenced under CAS 145915-63-5, blocks the de novo fibrillization process and disassembling preformed NM fibers. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.

Synonym(s):

Prionogenesis inhibitor, DAPH-12, Aβ42 Amyloidogenesis Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H19N3O4
CAS Number:
Molecular Weight:
389.40
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

orange

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable 4,5-dianilinophthalimide that antagonizes Aβ42 amyloidogenesis as well as yeast Sup35 N-terminal domain- (aa 1 -124; NM) mediated prionogenesis by blocking the de novo fibrillization process and disassembling preformed NM fibers. Treatment with DAPH-12 greatly reduces the ratio between prion-harboring ([PSI+]) and non-prion-harboring ([psi-]) yeast populations. DAPH-12 is also shown to effectively convert the [PSI+] phenotype into that of [psi-] in a dose-dependent manner, while exhibiting no activity against [RNQ+] prion-containing cells. While DAPH-12 is in general more effective than EGCG (Cat. No. 324880) in antagonizing the various forms of NM prions both in cell-free and cell-based system, these two reagents used in tandem can create a synergistic effect against antiprion activity.

Warning

Toxicity: Standard Handling (A)

Other Notes

Roberts, B.E., et al. 2009. Nat. Chem. Bio.5, 936.
Wang, H., et al. 2008. Proc. Natl. Acad. Sci. USA105, 7159.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service