Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

553003

Sigma-Aldrich

Raf1 Kinase Inhibitor I

InSolution, ≥97%

Synonym(s):

InSolution Raf1 Kinase Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H8Br2INO2
Molecular Weight:
520.94
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

clear orange

shipped in

wet ice

storage temp.

2-8°C

General description

A potent cRaf1 kinase inhibitor (IC50 = 9 nM). Shows ~100-fold selectivity for Raf kinase versus Cdk1, Cdk2, c-src, ERK2, MEK, p38, Tie2, VEGFR2 and c-fms. cRaf1 inhibitors are predicted to be broad spectrum anti-tumor agents.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
CRAFI kinase
Product competes with ATP.
Reversible: yes
Target IC50: 9 nM against cRAF1 kinase

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 10 mM (500 µg/96 µl) solution of Raf1 Inhibitor I (Cat. No. 553008) in DMSO.

Reconstitution

Following initial use, aliquot and refrigerate (4°C). Stock solutions are stable for up to 3 months at 4°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service