Skip to Content
MilliporeSigma

559274

S6K1 Inhibitor II, DG2

The S6K1 Inhibitor II, DG2 controls the biological activity of S6K1. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

S6K1 Inhibitor II, DG2, 3-Bromo-4-(4-(2-methoxyphenyl)piperazin-1-yl)-1H-pyrazolo[3,4-d]-pyrimidine

Sign Into View Organizational & Contract Pricing

Select a Size


About This Item

Empirical Formula (Hill Notation):
C16H17BrN6O
CAS Number:
Molecular Weight:
389.25
MDL number:
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL, clear, pale yellow

shipped in

ambient

storage temp.

2-8°C

SMILES string

BrC1=NNC2=C1C(N(CC3)CCN3C4=C(C=CC=C4)OC)=NC=N2

General description

A cell-permeable pyrazolopyrimidine compound that acts as a potent, ATP-competitive and reversible inhibitor of S6K1 (p70 ribosomal protein S6 kinase 1; IC50 = 9.1 nM) with ~2,400 reduced activity towards Akt (IC50 = 22 µM). Shown to completely block rpS6-Ser240/244 phosphorylation in L6 cells at 2.5 µM without inducing pAkt-Thr308 and Ser473, and display substantial selectivity in a 219-kinase panel (1 µM of DG2 in the presence of 10 µM of ATP; IC50 ≥50%; Clk1, Clk2, Dyrk1A, Dyrk3, Flt3, Flt3-D835Y, Flt4, GSK-3α/β, KDR, MAP4K4, Mink1, PKA, PRKG1, PRKG2, Ret-V804L, Ret-Y791F, RSK1, RSK3, RSK2, MSK2, MSK1 and p70S6K).

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Wang, B.T., et al. 2011. Proc. Natl. Acad. Sci. USA108, 15201.
Okuzumi, T., et al. 2009. Nat. Chem. Biol.5, 484.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zewen Jiang et al.
Cell metabolism, 33(9), 1836-1852 (2021-08-05)
With the increasing prevalence of type 2 diabetes and fatty liver disease, there is still an unmet need to better treat hyperglycemia and hyperlipidemia. Here, we identify isthmin-1 (Ism1) as an adipokine and one that has a dual role in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service