Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

559408

Sigma-Aldrich

SB 220025, Trihydrochloride

Synonym(s):

5-(2-Amino-4-pyrimidinyl)-4-(4-fluorophenyl)-1-(4-piperidinlyl)imidazole, p38 MAP Kinase Inhibitor XIV

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$190.00

$190.00


Please contact Customer Service for Availability


Select a Size

Change View
500 μG
$190.00

About This Item

UNSPSC Code:
41106305

$190.00


Please contact Customer Service for Availability

form

white solid

Quality Level

storage condition

OK to freeze

solubility

soluble (H2O (22 mg/ml))

storage temp.

−20°C

General description

A cell-permeable, potent, reversible, ATP-competitive, and specific inhibitor of human p38 MAP Kinase (IC50 = 60 nM). Binds to an extended pocket in the active site of the enzyme. Exhibits over 2000-fold greater selectivity for p38 MAPK over ERK (p42/p44 MAP kinase), 500-fold over PKA, 50-fold over PKC, and >1000-fold over EGFR. Also acts as a potent inhibitor of angiogenesis and as an inhibitor of LPS-induced TNF-α production.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service