The γ₄₀-Secretase Inhibitor I controls the biological activity of γ₄₀-Secretase. This small molecule/inhibitor is primarily used for Neuroscience applications.
Synonym(s):
γ4₀-Secretase Inhibitor I, t-3,5-DMC-IL-CHO, N- trans-3,5-Dimethoxycinnamoyl)-Ile-leucinal, t-3,5-DMC-IL-CHO, N-trans-3,5-Dimethoxycinnamoyl)-Ile-leucinal
A potent, cell-permeable, reversible γ-secretase inhibitor that preferentially appears to inhibit the secretion of Aβ1-40 (>90%) over Aβ1-42 (~15%). Aβtotal IC50 ~ 15 M; Aβ1-40 IC50 ~ 22 M; Aβ1-42 IC50 >50 M in CHO cells stable transfected with the cDNA encoding βAPP695.
A potent, cell-permeable, reversible inhibitor of γ-secretase that is reported to preferentially inhibit the secretion of Aβ1-40 (>90%) vs. Aβ1-42. IC50 = ~15 µM for Aβtotal; ~22 µM for Aβ1-40; and >50 µM for Aβ1-42 in CHO cells stably transfected with the cDNA encoding βAPP695. Reported to be about 10-fold more potent than Z-Val-Phe-CHO [MDL 28170 (Cat. No. 208722)].
Biochem/physiol Actions
Cell permeable: yes
Primary Target secretion of Aβ₁₋₄₀
Product does not compete with ATP.
Reversible: yes
Target IC50: ~ 15 µM for Aβtotal; ~ 22 µM for Aβ1-40; <50 µM for Aβ1-42 in CHO cells stable transfected with the cDNA encoding βAPP695
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.
Other Notes
Higaki, J.N., et al. 1999. J. Med. Chem. 42, 3889.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.