Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

606015

Sigma-Aldrich

Thymidylate Kinase Inhibitor, YMU1

The Thymidylate Kinase Inhibitor, YMU1 controls the biological activity of Thymidylate Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Thymidylate Kinase Inhibitor, YMU1, Ethyl 4-(2-(4,6-dimethyl-3-oxoisothiazolo[5,4-b]pyridin-2(3H)-yl)acetyl)piperazine-1-carboxylate, TMPK Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22N4O4S
CAS Number:
Molecular Weight:
378.45
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥99% (HPLC)

form

solid

potency

610 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 3.3 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

CC1=NC(SN(CC(N2CCN(CC2)C(OCC)=O)=O)C3=O)=C3C(C)=C1

General description

A cell-permeable, non-toxic pyridineisothiazolone compound that acts as a potent, reversible, and ATP-competitive inhibitor of human Thymidylate kinase (TMPK) (IC50 = 610 nM; Ki = 180 nM). Does not affect the activity of Thymidine kinase 1 (TK1) (IC50 >10 µM). Its action appears to involve blocking the interaction of Mg2+ with Asp15 in the catalytic domain. Shown to specifically reduce dTTP (deoxythymidine triphosphate) by 30 to 40% in HCT-116 p53-/- cells, without affecting dATP, dCTP, and dGTP levels. Significantly increases (3 to 35 fold) the sensitivity of a variety of tumor cell lines to doxorubicin (>Cat. No. 324380), and shrinks tumor size in HCT-116 p53-/- xenografted mouse model (doxorubicin, 1.25 mg/kg twice a week, i.p.; YMU1, 5 mg/kg thrice a week, i.p.).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
TMPK
Reversible: yes
Target Ki: 180 nM for TMPK

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C. Use only fresh DMSO for reconstitution.

Other Notes

Hu, C.M., et al. 2012. Cancer Cell22, 36.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service