69068
Ekapture Agarose
Sign Into View Organizational & Contract Pricing
Select a Size
1.5 ML
$172.00
10 ML
$876.00
About This Item
UNSPSC Code:
12352201
NACRES:
NA.32
Skip To
Quality Level
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
No data available
Flash Point(C)
No data available
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service