Key Documents
69439
Sigma-Aldrich
pET-11d DNA
Novagen′s pET-11a-d vectors carry an N-terminal T7-Tag sequence and BamH I cloning site. The pET-21a-d(+) series corresponds to pET-11a-d but incorporates several additional features.
Synonym(s):
cloning vector, expression vector, vector
Select a Size
10 μG
$340.00
$340.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
10 μG
$340.00
About This Item
UNSPSC Code:
41106610
NACRES:
NA.51
$340.00