Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

70573

Sigma-Aldrich

Clonables 2X Ligation Premix - Novagen

One solution for optimal ligation in 15 minutes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116133
Pricing and availability is not currently available.

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

dry ice
wet ice

storage temp.

−70°C

General description

The Clonables 2X Ligation Premix is a single solution containing optimized concentrations of the highest quality T4 DNA ligase, buffer, stabilizer, and cofactors needed for efficient ligation of any type of compatible DNA ends. The premix is tested for ligation of compatible 2- to 4-base cohesive DNA ends as well as blunt ends and single-base overhangs found on some PCR products. Under most conditionsexcellent ligation is achieved in only 15 minutes; with T/A or U/A overhangs more colonies can be obtained by incubating up to two hours. Although maximal efficiencies are obtained using NovaBlue Singles Competent Cells, this reagent is compatible with transformation of any type of chemically competent cells provided by the user. The high performance, minimal pipetting requirement, and single-addition format make the premix suitable for high-throughput applications.

Packaging

Plastic ampoule

Warning

Toxicity: Irritant

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service