70597
Introductory pETBlue-1 AccepTor Vector Kit - Novagen
Select a Size
Select a Size
About This Item
manufacturer/tradename
Novagen®
Quality Level
storage condition
OK to freeze
shipped in
dry ice
storage temp.
−70°C
General description
Features and Benefits
- No restriction enzymes or special primers
- Direct ligation of PCR product with vector
- Compatible with polymerases that leave single 3´-dA overhangs
- Blue/white screening with pSTBlue-1 or pETBlue-1 vectors
- Simple protocol takes as little as 40 minutes from PCR product to plating transformants
Packaging
Components
- 0.5 µg- pETBlue-1 AccepTor Vector
- 10 µl- AccepTor Vector Positive Control Insert
- 55 µl- Clonables 2X Ligation Mix
- 1.5 ml- Nuclease-free Water
- 11 × 50 µl- NovaBlue Singles Competent Cells
- 3 × 2 ml- SOC Medium
- 10 µl- Test Plasmid
- 0.2 ml- Tuner (DE3) pLacI Competent Cells
Warning
Legal Information
Disclaimer
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service