Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABS12

Sigma-Aldrich

Anti-Dia 1 Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

conjugate

unconjugated

Quality Level

antibody form

affinity purified immunoglobulin

clone

polyclonal

purified by

affinity chromatography

species reactivity

rabbit, mouse, human

concentration

(Please refer to lot specific datasheet.)

technique(s)

immunocytochemistry: suitable
western blot: suitable

UniProt accession no.

target post-translational modification

unmodified

General description

KLH-conjugated linear peptide corresponding to the FH2 domain of human Dia 1.

Application

Epitope: FH2 Domain

Quality

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected Dia 1 in 10 µg of A431 cell lysate.

Physical properties

~160 kDa observed

Physical form

Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service