Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AX1699A

Supelco

Water/Methanol Standard

5000 ppm (0.5%), for use with AQUASTAR®

Synonym(s):

Water/Methanol Standard, Karl Fischer

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH4O
Molecular Weight:
32.04
UNSPSC Code:
50202301
EC Index Number:
200-659-6

form

liquid

Quality Level

concentration

5000 ppm (0.5%)

compatibility

for use with AQUASTAR®

shipped in

ambient

storage temp.

room temp

General description

5000 ppm 0.5% AQUASTAR®

Application

AX1699A-6 is packaged in 38/430 poly coated glass bottles

Analysis Note

Expiration date: Exp 24 months from mfg date
Color (APHA): 10 max
Form: Clear liquid
Water (H2O): 4.98-5.02mg/mL

Legal Information

AQUASTAR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service