Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BX1410

Supelco

Bromophenol blue

pH indicator, ACS, GR ACS

Synonym(s):

Bromophenol Blue, 3`,3``,5`,5``-tetrabromo-m-cresolsulfonephthalein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H10Br4O5S
CAS Number:
Molecular Weight:
669.96
MDL number:
UNSPSC Code:
12171502
EC Index Number:
204-086-2

Product Name

Bromophenol Blue, Meets ACS Specifications GR ACS

form

powder

color

light red to purple

visual transition interval

3.0-4.6, yellow to blue

mp

270-273 °C (decomposition)

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C19H10Br4O5S/c20-12-5-9(6-13(21)17(12)24)19(10-7-14(22)18(25)15(23)8-10)11-3-1-2-4-16(11)29(26,27)28-19/h1-8,24-25H

InChI key

UDSAIICHUKSCKT-UHFFFAOYSA-N

Application

Meets ACS Specifications GR ACS

Analysis Note

Clarity of solutionTo pass testColorLight red to purpleFormPowderVisible spectrum, max. absorbance0.60AU minVisible spectrum, max. wavelength600-605nmVisual transition (from pH 3.0 to pH 4.6)Yellow to Blue

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service