Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BX1636

Supelco

Buffer pH 10.0 Clear

Synonym(s):

buffer solutions

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

analytical standard

Quality Level

Agency

traceable to NIST

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

shipped in

ambient

storage temp.

15-25°C

Application

This pH 10.0 buffer reference solution is suitable for use in daily analytical procedures involving the calibration of pH meters and maintaining pH of solutions.

Features and Benefits

  • Traceable to NIST standards
  • A ready-to-use solution available in a sealed bottle with convenient packaging to protect against contamination
  • Accompanied with a certificate of analysis (CoA) for an effective audit
  • Accurate up to +/- 0.01 at 25 °C
  • The expiry date of the product can be found in the CoA
  • Uncertainty value calculated includes for the duration of transportation and storage as well

Specifications

Tolerance: ±0.01

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service