MilliporeSigma
All Photos(3)

EC111

Sigma-Aldrich

GenElute-E Tissue Stabilizer

greener alternative

feature

Intended use (For stabilization of tissue samples prior to DNA or RNA purification)

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

greener alternative category

storage temp.

room temp

General description

For storage of human or animal tissue samples and protection of DNA and RNA against degradation during storage. GenElute-E Tissue Stabilizer provides non-toxic stabilization of DNA or RNA isolated from tissue. Not intended for use with bacteria or cultured cells.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is aligned with "Waste prevention" and "Designing safer chemicals". Click here for more information.

Features and Benefits

Non-toxic stabilization of DNA or RNA isolated from tissue

Storage and Stability

Use within 12 months of receipt.

Other Notes

We believe that green chemistry will contribute to a better tomorrow. With a growing portfolio of greener alternatives, there are now more choices to reduce the ecological impact of your research while still delivering quality and efficacy so your results are not compromised. GenElute-E kits adhere to the principles of SMASH Packaging, our global initiative to improve the sustainability through the reduction of packaging materials, the increased use of sustainable alternatives, and improved recyclability. Packaging is FCS-marked for sustainable forestry practices and produced from greater than 70% recycled content.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Quality

Enter Lot Number to search for Certificate of Quality (COQ).

Product Information Sheet

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service