All Photos(3)
EC9999
Sigma-Aldrich
GenElute-E Single Spin Cap Puncher
greener alternative
feature
Intended use (For use with GenElute-E Single Spin purification columns)
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.
greener alternative category
Compare Similar Items
View Full Comparison
1 of 4
This Item
CLS431483CLS431490CLS431489
feature
Intended use (For use with GenElute-E Single Spin purification columns)
feature
-
feature
-
feature
-
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.
greener alternative product characteristics
-
greener alternative product characteristics
-
greener alternative product characteristics
-
greener alternative category
Aligned
greener alternative category
-
greener alternative category
-
greener alternative category
-
General description
GenElute-E purification kits provide a simple alternative for DNA purification. Compared with traditional methods, GenElute-E offers:
  • Isolation of DNA in under an hour with minimal sample handling
  • Better purity, leading to improved performance in PCR and other applications
  • Significant reduction in plastic and hazardous chemical waste
Traditional silica-based, bind-wash-elute purification kits require multiple wash steps to remove impurities from the spin columns. These steps increase the risk of cross contamination, subject the DNA to centrifugation sheering forces, and introduce chaotropic salts that can carry over into the final sample, inhibiting downstream applications.
GenElute-E kits employ size exclusion negative chromatography to separate large nucleic acid molecules from smaller protein, lipid, and ionic components in cell, tissue, blood, and other samples. Single-spin columns efficiently absorb and retain cellular debris and sample contaminants while allowing nucleic acids to pass through, reducing the number of steps and plastic materials required for purification. This novel method for high-quality purification is made possible by our innovative SmartLyse® Protease, which enables fast and efficient lysis of a wide range of sample types.
Isolate nucleic acid in a fraction of time compared to traditional silica bind-wash-elute procedures. Simply mix sample with lysis components and incubate according to kit protocol, pipette sample onto the spin column, and centrifuge directly into a collection tube. Cellular debris and contaminants remain bound in the column to be discarded, while purified DNA is ready to be used in downstream applications or stored.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is aligned with "Waste prevention" and "Designing safer chemicals". Click here for more information.
Application
The GenElute-E Cap Puncher is designed to work with the GenElute-E line of DNA and RNA purification kits. When pressed onto the lid of the single-spin column, the cap puncher creates a small opening through which a sample can be pipetted directly onto the column. This reduces the amount of handling by eliminating the need for cap removal, and lessens the chance of cross contamination when handling multiple samples.For full instructions, please reference any of the GenElute-E protocols.
Other Notes
We believe that green chemistry will contribute to a better tomorrow. With a growing portfolio of greener alternatives, there are now more choices to reduce the ecological impact of your research while still delivering quality and efficacy so your results are not compromised.  GenElute-E purification kits avoid the use of hazardous wash buffers typically employed in silica-based purification kits, significantly reducing chemical waste. The single-spin protocol also limits the number of collection steps, reducing plastic waste. GenElute-E is designated a Greener Alternative Product based on the "Prevention" and "Designing Safer Chemicals" qualifications of The 12 Principles of Green Chemistry.GenElute-E kits also adhere to the principles of SMASH Packaging, our global initiative to improve the sustainability through the reduction of packaging materials, the increased use of sustainable alternatives, and improved recyclability. Packaging is FCS-marked for sustainable forestry practices and produced from greater than 70% recycled content.
Legal Information
GenElute is a trademark of Sigma-Aldrich Co. LLC
SmartLyse is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificate of Analysis
Please enter the Product Number and Lot Number.
Certificate of Quality
Please enter the Product Number and Lot Number.
Product Information Sheet