Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EX0272

Supelco

Specially Denatured Alcohol 3A

190 Proof

Synonym(s):

ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352104

Quality Level

form

liquid

concentration

190 proof in alcohol (denatured)

transition temp

flash point 12 °C (Information refers to the main component.)

shipped in

ambient

storage temp.

room temp

Application

190 Proof

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - Information refers to the main ingredient.

Flash Point(C)

12 °C - Information refers to the main ingredient.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service