MilliporeSigma
All Photos(1)

Documents

GF340

Sigma-Aldrich

IL-9,
Human Recombinant Animal Free

Sign Into View Organizational & Contract Pricing

Select a Size

Synonym(s):
Interleukin-9, IL9
eCl@ss:
32160405

impurities

<0.1 μg/mg protein endotoxin (<1 EU/μg)

Quality Level

UniProt accession no.

shipped in

dry ice

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF325GF327GF331
UniProt accession no.

P15248

UniProt accession no.

P15248

UniProt accession no.

P40933

UniProt accession no.

P01584

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

General description

IL-9 is an immunoregulatory cytokine produced by IL-2 activated Th2 lymphocytes. IL-9 enhances the proliferation of T lymphocytes, mast cells, erthroid precursor cells and megakaryoblastic leukemia cell lines. Over-expression of IL-9 has been implicated in the pathogenesis of anaplastic lymphoma and Hodgkin’s disease. Whereas murine IL-9 can function on human cells, human IL-9 is inactive on mouse cells.
Recombinant human IL-9 is a 14.0 kDa protein of 127 amino acid residues including 10 cysteine residues which are fully conserved between the human and murine proteins.
Product Source: Protein is expressed in E.coli.
The protein is manufactured without the use of any animal products/reagents and is considered Animal-Free and can be used in protocols where possible transmisson of animal viruses and contamination must be avoided.

Application

Research Category
Stem Cell Research
Research Sub Category
Growth Factors & Receptors

Quality

Determined by its ability to stimulate the proliferation of human MO7e cells. The expected ED50 is ≤ 0.2 ng/ml, corresponding to a specific activity of ≥ 5 x 10^6 units/mg.

Physical form

Product is presented without additives and is filtered through a 0.2 micron filter before lyophilization.

Storage and Stability

Store at -20°C for up to 4 months from date of receipt Centrifuge the vial prior to opening.
Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein and store in working aliquots at -20°C.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service