Skip to Content
MilliporeSigma

HTS070RTA

Sigma-Aldrich

Ready-to-Assay ETA Endothelin Receptor Frozen Cells

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$1,070.00

$1,070.00


Please contact Customer Service for Availability

Select a Size

Change View
1 KIT
$1,070.00

About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.76

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

$1,070.00


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

description

Cell line type: GPCR Cell Lines
GPCR Class A
protein target ETA

Quality Level

technique(s)

calcium flux assay: suitable

detection method

fluorometric

General description

Millipore′s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.

The endothelin family of peptides are potent vasoconstrictors synthesized by endothelial cells in both constitutive and inducible pathways. The biological actions of endothelins are mediated by two GPCRs, ETA and ETB. ETA is the predominant receptor on smooth muscle cells and thus is the primary mediator of the vasoconstrictor activity of endothelins in vivo. Endothelin receptors are also found in the epithelium and central nervous system (Davenport, 2002). Millipore′s cloned human ETA-expressing cell line is made in the Chem-1 host, which supports high levels of recombinant ETA expression on the cell surface for functional detection via the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists, and modulators at ETA .

Components

Pack contains 2 vials of mycoplasma-free cells, 1 ml per vial. Fifty (50) mL of Media Component.

Disclaimer

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges {HCompany} to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service