Skip to Content
MilliporeSigma

MABC1717

Sigma-Aldrich

Anti-Tn mIgM (STn-PS A1), clone KT-IgM-8

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$242.00
100 μL
$619.00

$242.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$242.00
100 μL
$619.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

$242.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

antibody form

purified antibody

clone

Kt-IgM-8
monoclonal

purified by

(Ligatrap)

species reactivity

human, human, mouse

concentration

(Please refer to lot specific datasheet.)

technique(s)

ELISA: suitable
flow cytometry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgM

suitability

suitable for immunoassay

General description

Semi-synthetic Tn-PS A1 (Tn= D-GalpNAc)

Specificity

Clone KT-IgM-8 is a mouse monoclonal antibody that detects Thomsen-nouveau (Tn) polysaccharide A1 monovalently, including in human tumor cells.

Application

Affects Function Analysis: A representative lot showed high complement-dependent cytotoxicity (CDC) activity. (Trabbic, K.R., et. al. (2018). Cancer Immunol Immunother. 67(9):1437-1447).

Flow Cytometry Analysis: A representative lot detected Tn-antigen in Flow Cytometry applications (Trabbic, K.R., et. al. (2018). Cancer Immunol Immunother. 67(9):1437-1447).

Enzyme Immunoassay Analysis: A representative lot detected Tn-antigen in ELISA applications (Trabbic, K.R., et. al. (2018). Cancer Immunol Immunother. 67(9):1437-1447).

Quality

Evaluated by Flow Cytometry in MCF-7 cells.

Flow Cytometry Analysis: 1 µg of this antibody detected Tn-antigen in one million MCF-7 cells.

Physical form

Purified mouse monoclonal antibody IgM in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service