Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF2187

Sigma-Aldrich

Anti-Zika Virus NS1 protein Antibody, clone 110

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

antibody form

purified antibody

clone

110
monoclonal

purified by

using protein G

species reactivity

mouse, virus

concentration

(Please refer to lot specific datasheet.)

technique(s)

ELISA: suitable
flow cytometry: suitable
western blot: suitable

isotype

IgG2aκ

General description

Recombinant Zika virus NS1 protein expressed in mammalian cells.

Specificity

Clone 110 is a mouse monoclonal antibody that specifically detects Zika virus NS1 protein.

Application

Flow Cytometry Analysis: A representative lot detected Zika Virus NS1 protein in Flow Cytometry applications (Bosch, I., et. al. (2017). Sci Transl Med. 9(409); eaan1589).

Enzyme Immunoassay (ELISA) Analysis: A representative lot detected Zika Virus NS1 protein in ELISA applications (Bosch, I., et. al. (2017). Sci. Transl. Med. 9(409); eaan1589).

Quality

Evaluated by Western Blotting with His-tagged recombinant Zika Virus NS1 protein.

Western Blotting Analysis: A 1:250 dilution of this antibody detected His-tagged recombinant Zika Virus NS1 protein.

Physical properties

~47 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Virus (Zika virus).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service