Skip to Content
MilliporeSigma

MABF2297

Sigma-Aldrich

Anti-IL-12 p70 Antibody, clone 2F7

clone 2F7

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

MABF2297-25UG
$238.00
MABF2297-100UG
$611.00

$238.00


Please contact Customer Service for Availability

Select a Size

Change View
MABF2297-25UG
$238.00
MABF2297-100UG
$611.00

About This Item

UNSPSC Code:
12352203

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

$238.00


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

clone

2F7, monoclonal

Quality Level

isotype

IgG1κ

Specificity

Clone 2F7 is a mouse monoclonal antibody that detects human Interleukin-12 p70.

Immunogen

His-tagged full-length recombinant human Interleukin-12A.

Reconstitution

1 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Stable for 1 year at +2°C to +8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service