Skip to Content
MilliporeSigma

MABN1866

Sigma-Aldrich

Anti-ANO6/TMEM16F Antibody, clone N429/19

clone N429/19

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$255.00
100 μL
$655.00

$255.00


Please contact Customer Service for Availability


About This Item

UNSPSC Code:
12352203

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

clone

N429/19, monoclonal

Quality Level

isotype

IgG2bκ

Immunogen

Recombinant fragment corresponding to the first 294 amino acids from the N-terminal, cytoplasmic domain of human Anoctamin-6 (ANO6).

Biochem/physiol Actions

Clone N429/19 is a mouse monoclonal antibody that detects Anoctamin-6 (TMEM16F) . It targets an epitope within the N-terminal cytoplasmic domain.

Preparation Note

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Stable for 1 year at +2°C to +8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service