Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABN2393

Sigma-Aldrich

Anti-TRAAK, clone N445_27

new
Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
Pricing and availability is not currently available.

antibody form

purified antibody

antibody product type

primary antibodies

clone

N445/27
monoclonal

mol wt

(43.05 kDa calculated.)

purified by

using protein G

species reactivity

mouse, rat

isotype

IgG2aκ

Protein ID accession no.

UniProt accession no.

Specificity

Clone N445/27 is a mouse monoclonal antibody that detects Potassium channel subfamily K member 4 (TRAAK). It targets an epitope within the C-terminal, cytoplasmic domain.

Application

Anti-TRAAK, clone N445/27, Cat. No. MABN2393, is a mouse monoclonal antibody that detects Potassium channel subfamily K member 4 (TRAAK) and has been tested for use in Immunohistochemistry (Paraffin).
Immunohistochemistry (Paraffin)
Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution from a representative lot detected TRAAK in rat brain tissue sections.

Features and Benefits

A recombinant fragment corresponding to 143 amino acids from the C-terminal, cytoplasmic domain of mouse Potassium channel subfamily K member 4 (TRAAK).

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Other Notes

Potassium channel subfamily K member 4 (UniProt: O88454; also known as TWIK-related arachidonic acid-stimulated potassium channel protein, TRAAK) is encoded by the Kcnk4 (also known as Traak) gene (Gene ID: 16528) in murine species. TRAAK is a multi-pass membrane protein that is expressed in brain, spinal cord, and eye and serves as a voltage-insensitive potassium channel. It is a mechano-activated channel that senses shear stress and negative membrane pressure. It generates progressive and non-inactivating current when negative pressure is applied to the plasma membrane. The channel opening is triggered by mechanical forces that deform the membrane. Channel opening is also triggered by raising the intracellular pH to basic levels. TRAAK channel is shown to be inactive at 24C, but raising the temperature to 37C or higher increases the frequency of channel opening. It plays a role in the sensory perception of pain caused by heat. The C-terminal of TRAAK has a charged cluster that is important in maintaining the mechanosensitive properties of the channel. TRAAK channels are reported to be gradually and reversibly activated by polyunsaturated fatty acids (PUFA) including arachidonic acid that potentiates TRAAK background currents by 5- to 20-fold. (Ref.: Noël, J., et al. (2011). Channels (Austin). 5(5); 402-409).
Evaluated by Immunohistochemistry (Paraffin) in mouse brain tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution of this antibody detected TRAAK in mouse brain tissue sections.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service