Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS2175

Sigma-Aldrich

Anti-FOLR1 Antibody, clone LK26

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$242.00
100 μG
$619.00

$242.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$242.00
100 μG
$619.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$242.00


Please contact Customer Service for Availability

Request a Bulk Order

antibody form

purified antibody

clone

LK26
monoclonal

purified by

using protein G

species reactivity

human, human, mouse

concentration

(Please refer to lot specific datasheet.)

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG2aκ

UniProt accession no.

General description

Gestational choriocarcinoma cells (Luc-75(c))

Specificity

Clone LK26 is a mouse monoclonal antibody that detects human Folare receptor 1.
Clone LK26 is a mouse monoclonal antibody that detects human Folate receptor 1.

Application

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected FOLR1 in human placenta tissue sections.

Immunohistochemistry (Paraffin) Analysis: A representative lot detected FOLR1 in Immunohistochemistry applications (Garin-Chesa, P., et. al. (1993). Am J Pathol. 142(2):557-67).

Immunohistochemistry Analysis: A representative lot detected FOLR1 in Immunohistochemistry applications (Rettig, W.J., et. al. (1985). Int J Cancer. 35(4):469-75).

Immunoprecipitation Analysis: A representative lot immunoprecipitated FOLR1 in Immunoprecipitation applications (Rettig, W.J., et. al. (1985). Int J Cancer. 35(4):469-75).

Quality

Evaluated by Immunohistochemistry (Paraffin) in human fallopian tube tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected FOLR1 in human fallopian tube tissue sections.

Physical properties

29.82 kDa calculated.

Physical form

Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service