R04561
SAF Fixative
Harleco®, Acetate Buffered Formalin with Triton
Synonym(s):
fixative
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116113
Recommended Products
Quality Level
manufacturer/tradename
Harleco®
color
colorless
application(s)
diagnostic assay manufacturing
hematology
histology
shipped in
ambient
storage temp.
15-25°C
General description
HARLECO® Acetate Buffered Formalin with Triton
Application
HARLECO® Acetate Buffered Formalin with Triton
Legal Information
HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service