Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TX0255

Sigma-Aldrich

Tetraethylammonium Bromide

Synonym(s):

Tetraethylammonium Bromide, TEAB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)4NBr
CAS Number:
MDL number:
UNSPSC Code:
12352107
EC Index Number:
200-769-4

Assay

≥99%

form

crystals

pH

6.5 (20 °C, 100 g/L in H2O)

mp

>285 °C

solubility

2795 g/L

density

1.397 g/cm3 at 20  °C

shipped in

ambient

storage temp.

room temp

InChI

1S/C8H20N.BrH/c1-5-9(6-2,7-3)8-4;/h5-8H2,1-4H3;1H/q+1;/p-1

InChI key

HWCKGOZZJDHMNC-UHFFFAOYSA-M

Analysis Note

Assay: 99% min
Color: White
Form: Crystals
Infrared Spectrum: Conforms to standard

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

361.4 °F - closed cup

Flash Point(C)

183 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service