MilliporeSigma
All Photos(1)

W2660

SAFC

1,2-Distearoyl-sn-glycero-3-phosphocholine

DSPC, GMP, powder

Synonym(s):
PC(18:0/18:0), 18PC, 3-sn-Phosphatidylcholine, 1,2-distearoyl, 1,2-Dioctadecanoyl-sn-glycero-3-phosphocholine, DSPC, PC, 18:0 PC, Phospholipid, L-β,γ-Distearoyl-α-lecithin, L-α-Phosphatidylcholine, distearoyl
Empirical Formula (Hill Notation):
C44H88NO8P
CAS Number:
Molecular Weight:
790.15
Beilstein:
3923978
MDL number:

Quality Level

biological source

synthetic

grade

GMP

form

powder

packaging

pkg of 1 g
pkg of 10 g
pkg of 100 g
pkg of 1000 g

transition temp

transition temp 55 °C

application(s)

pharmaceutical

storage temp.

−20°C (−15°C to −25°C)

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O)=O

InChI

1S/C44H88NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-43(46)50-40-42(41-52-54(48,49)51-39-38-45(3,4)5)53-44(47)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h42H,6-41H2,1-5H3/t42-/m1/s1

InChI key

NRJAVPSFFCBXDT-HUESYALOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A GMP-scalable process which yields a high-quality lipid is essential to lipidic drug product development and manufacturing. We are highly specialized in synthesizing top-quality GMP lipids for pharmaceutical applications and gene therapy, focusing on tailored manufacturing to meet your specific needs. We also offer an off-the-shelf catalog with optimized stability, solubility, and handling characteristics.
Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
Non-pyrogenic, well-defined liposomes, loaded with a molecule of choice, are formed by a single hydration step.

Application

DSPC is used as a structural lipid in the preparation of lipid-based systems like liposomes and lipid nanoparticles (LNPs).
This product is GMP grade and suitable for divers formulation applications ranging from lipid nanoparticle and liposomes to microbubbles for ultrasound contrast agents.

Packaging

Amber glass screw cap bottle, bubble wrap outer bag

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Quality

Enter Lot Number to search for Certificate of Quality (COQ).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service