Select a Size
$254.00
Select a Size
About This Item
$254.00
Recommended Products
description
Powered by Eupheria Biotech
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
AGTTGAAAGATCTGCACGTGATAGACATGAAGACATCCATTCTTCTGCCAGCAGAAATACCAACAACCAGTTCCAGTTCTCAGCATTTCAACCTGAACTTCACCATCACCAACCTACCCTATTCCCAAGACATAGCCCAGCCAAGCACCACCAAATACCAGCAAACCAAAAGAAGTATTGAAAATGCGCTCAATCAGCTTTTCAGAAACAGCAGCATCAAGAGTTACTTTTCTGACTGTCAAGTATTAGCCTTCAGGTCTGTCTCCAACAACAACAACCACACAGGGGTAGATTCCCTATGTAACTTCTCACCATTGGCTCGGCGAGTAGACAGAGTTGCTATCTATGAGGAATTCCTTCGAATGACGCACAATGGTACCCAGTTGCTGAACTTCACACTGGATAGGAAGAGTGTCTTTGTGGATGGGTATTCTCAAAACAGAGATGATGATGTGATGAAGAATTCTGGTCTTCCTTTCTGGGCTATCATTCTCATCTGCTTGGCGGTACT
Ensembl | mouse accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
mouse ... MUC16(73732)
Related Categories
1 of 4
This Item | EMU060891 | EMU049511 | EMU050641 |
---|---|---|---|
product line MISSION® | product line MISSION® | product line MISSION® | product line MISSION® |
esiRNA cDNA target sequence AGTTGAAAGATCTGCACGTGATAGACATGAAGACATCCATTCTTCTGCCAGCAGAAATACCAACAACCAGTTCCAGTTCTCAGCATTTCAACCTGAACTTCACCATCACCAACCTACCCTATTCCCAAGACATAGCCCAGCCAAGCACCACCAAATACCAGCAAACCAAAAGAAGTATTGAAAATGCGCTCAATCAGCTTTTCAGAAACAGCAGCATCAAGAGTTACTTTTCTGACTGTCAAGTATTAGCCTTCAGGTCTGTCTCCAACAACAACAACCACACAGGGGTAGATTCCCTATGTAACTTCTCACCATTGGCTCGGCGAGTAGACAGAGTTGCTATCTATGAGGAATTCCTTCGAATGACGCACAATGGTACCCAGTTGCTGAACTTCACACTGGATAGGAAGAGTGTCTTTGTGGATGGGTATTCTCAAAACAGAGATGATGATGTGATGAAGAATTCTGGTCTTCCTTTCTGGGCTATCATTCTCATCTGCTTGGCGGTACT | esiRNA cDNA target sequence AGGGATGAAGAGAGCCCCTATGCTGCCTCTCTTTATCATAGTTAGTGACTGACCAGCGAACTGATCCAGATCACAGAATGGCTGGACCCTAAGAATCCCGCCGGCTCTGGTTCAGGGGCAGCGCTGCCCCTGGACTCCCGTAGCGCACCTTCGAGCTGCAGTTAAGTAGCCCTACTGCAGCTTGCCTGAAGTGATCAGCTCCCGGCTCTCTATAGTTAAGTCTTATTACGGCATGATAGCAAAACTGTATTTATTCTAACGCCTTTCACTTTTGTAGATGGTGGTAGTTTTAGACCGGCTTTTGTAGATACAGAAAGCATCAGAGACTCTATATATTTAAGCCAAAAGCTATGTTTATTTCCTGATGAGATCCTAAATGTGTGGAAGACATGATGACTGTGG | esiRNA cDNA target sequence CGGAAGGAGAGAATGACAGCTTCGGGGAAGCAGAGTATGCTGCCATCAACTCCATGTTGGATCAGATCAACTCTTGTCTGGACCACCTGGAGGAGAAGAATGACCACCTCCATGCCCGCCTCCAGGAGCTGCTGGAGTCGAACCGGCAGACGCGCCTGGAATTCCAGCAACAGCTTGGGGAGGCCCCTGGCGATGCCAGCCCCTAGGTGCCGGGAGCCCCCATCCAGGCCCTACCCTCACCTCTCTAGGCCATGTTCTGGCCTGGGTAGATACTACTTGGCTTAGACACCATCTCGGGTACTGGCCTCCAGATCCTAGTGGGTCTACCAGCCTGGACCAGTCCCCATTCACTGCCCATCACCCTTCCTGGAGTCAGGTGCAATCCTACAGTTCTCCCACTTGTCTGTCTTCTTTCCCCTCCATCC | esiRNA cDNA target sequence CTATGCCTGCACCTCTCACACGCAGGGCCTGAGTCCAGGCTGCTATGACACATACAATGCGGACATTGACTGCCAGTGGATCGACATAACTGATGTGCAGCCTGGGAACTACATCCTCAAGGTGCACGTGAACCCCAAGTACATTGTTCTGGAGTCTGACTTCACCAACAACGTGGTGAGATGCAACATCCACTACACAGGTCGCTACGTTTCTACAACAAACTGCAAAATCGTCCAGTCCTGATCTGAGTGGACAGAGCCCCCCTTGTCTCTCCTTGAAAAGCAGGACCTGCTTCCAGGCAGCCTCCCCCATGAGGCTCAGCCGCCTAGCCCAAGGCAAAGGCAGCTGGCCGCTGGGGCATCCCTCCCTGCCGGCCTCAGGGAGTGAACATGGACCCAAACCACAGGGATTCTGGATGCCAGACCCTGTCTTGT |
Ensembl | mouse accession no. | Ensembl | mouse accession no. | Ensembl | mouse accession no. | Ensembl | mouse accession no. |
Gene Information mouse ... MUC16(73732) | Gene Information mouse ... Lysmd2(70082) | Gene Information mouse ... 1110008P14RIK(73737), 1110008P14Rik(73737) | Gene Information mouse ... LOXL1(16949), Loxl1(16949) |
storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
form lyophilized powder | form lyophilized powder | form lyophilized powder | form lyophilized powder |
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service