Skip to Content
MilliporeSigma

Skip To

VIV102005SB3

SynVivo SynBBB 3D blood brain barrier model chip, IMN2 radial configuration

case of 3 ea

Synonym(s):

Synvivo 102005-SB3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
3 ea
Please contact Customer Service for Availability
$467.00

About This Item

NACRES:
NA.84
UNSPSC Code:
60104506

$467.00


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

case of 3 ea

manufacturer/tradename

SynVivo 102005-SB3

storage temp.

room temp

General description

SynBBB Blood Brain Barrier Model Chip - Basic configuration - Barrier type: Slits, Slit width: 3 μm, Distance between slits: 50 μm, Travel/barrier width: 50 μm, Outer Channel width: 200 μm, Device depth: 100 μm.This chip is used to create the SynBBB model in monoculture, co or tri-culture configurations.

Other Notes

Link to Publications application text


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library