Skip to Content
MilliporeSigma

Skip To

VIV102016STO3

SynVivo SynTox 3D Toxicology model chip, IMN2 radial configuration

case of 3 ea

Synonym(s):

Synvivo 102016-STO3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
3 ea
Please contact Customer Service for Availability
$467.00

About This Item

NACRES:
NB.35
UNSPSC Code:
60104506

$467.00


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

case of 3 ea

manufacturer/tradename

SynVivo 102016-STO3

storage temp.

room temp

General description

SynTox 3D toxicology model chip- Barrier type: Slits, Slit width: 2 μm, Distance between slits: 50 μm, Travel/barrier width: 50 μm, Outer Channel width: 200 μm, Device depth: 100 μm. This chip is used to create the SynTox model in monoculture, co or tri-culture configurations.
SynTox is the only commercially available 3D toxicology model with real-time optical monitoring and multi-compartment, multi-cellular architecture and low reagent requirements. Other benefits of this platform are:
  • Physiologically realistic morphological, fluidic and 3D cellular conditions
  • Universal platform with architecture specific of desired organ
  • Significant reduction in cost and timeRobust and easy to use protocols
  • Compatible with standard analytical instruments for both on chip and off chip assays including omic methodologies for systems biology and bioinformatic analysis

Other Notes

LInk to Publications application text


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
VIV102016STO304065271239111