Skip to Content
MilliporeSigma

VIV1050013SR3

SynVivo SynRAM 3D Inflammation model chip, SMN2 microvascular network configuration

case of 3 ea

Synonym(s):

Synvivo 105001-3

Sign In to View Organizational & Contract Pricing.

Select a Size

3 EA

$629.00

$629.00


Please contact Customer Service for Availability


About This Item

NACRES:
NA.84
UNSPSC Code:
60104506

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

SynVivo SynRAM 3D Inflammation model chip, SMN2 microvascular network configuration, case of 3 ea

packaging

case of 3 ea

manufacturer/tradename

SynVivo 105001-SR3

storage temp.

room temp

General description

SynRAm inflammation model co-culture chip- Microvascular design Barrier type: Pillars, Distance between pillars: 50 μm, Pillar Diameter: 10 μm, Barrier height: 3 μm, Travel/barrier width: 50 μm, Device depth: 100 μm. The SynRAM 3D Inflammation Model has been developed to study the entire inflammation pathway in a realistic and dynamic environment.

Other Notes

LInk to Publications application text


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service