Skip to Content
MilliporeSigma

VIV105015STU3

SynVivo SynTumour 3D cancer model chip, microvascular network configuration, 8 um pillar

case of 3 ea

Synonym(s):

Synvivo 105015-STU3

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

NACRES:
NB.35
UNSPSC Code:
60104506
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

packaging

case of 3 ea

manufacturer/tradename

SynVivo 105015-STU3

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

SynTumor cancer model chip- Microvascular design-Barrier type: Pillars, Distance between pillars: 50 μm, Pillar Diameter: 10 μm, Barrier height: 8 μm, Travel/barrier width: 50 μm, Device depth: 100 μm. SynTumor is a 3D tissue model for real-time visualization and quantitative assessment of cell-cell and cell-drug interactions in a physiologically and morphologically realistic tumor microenvironment.

Other Notes

LInk to Publications application text


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service