MilliporeSigma
All Photos(1)

33350-U

Supelco

HMDS, Derivatization Grade

Synonym(s):
Hexamethyldisilazane
CAS Number:
EC Number:

grade

for GC derivatization

form

liquid

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

packaging

pkg of 1 × 30 mL

InChI

1S/C6H19NSi2/c1-8(2,3)7-9(4,5)6/h7H,1-6H3

InChI key

FFUAGWLWBBFQJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for schiff base, trimethylsilyl and trimethylsilyl ether.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service