Products
CA
EN
Products
Applications
Products
Services
Support
Home
Search Results
1,1,1,3,3,3-Hexamethyldisilazane
(2)
1,1,1,3,3,3-Hexamethyldisilazane
Synonyms:
1,1,1,3,3,3-Hexamethyldisilazane
Empirical Formula (Hill Notation):
C
6
H
19
NSi
2
CAS Number:
999-97-3
Molecular Weight:
161.39
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
8.04324
1,1,1,3,3,3-Hexamethyldisilazane, for synthesis
Expand
View Pricing
1.12186
1,1,1,3,3,3-Hexamethyldisilazane, for gas chromatography
Expand
View Pricing