Skip to Content
MilliporeSigma
Erastin ≥98% (HPLC)

Erastin

Erastin ≥98% (HPLC)
Synonym(s):
Erastin, 2-[1-[4-[2-(4-Chlorophenoxy)acetyl]-1-piperazinyl]ethyl]-3-(2-ethoxyphenyl)-4(3H)-Quinazolinone
Empirical Formula (Hill Notation):
C30H31ClN4O4
CAS Number:
Molecular Weight:
547.04

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-2 of 2