MilliporeSigma
Hydrochloric Acid 31.5-35.5%

Hydrochloric acid

Hydrochloric Acid 31.5-35.5%
Synonyms:
Hydrochloric acid, Hydrochloric acid solution, Hydrogen chloride solution
Linear Formula:
HCl
CAS Number:
Molecular Weight:
36.46

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Showing 1-16 of 16
Page 1 of 1
Page 1 of 1