MilliporeSigma
Hydrochloric acid 25% for analysis EMSURE®

Hydrochloric acid 25%

Hydrochloric acid 25% for analysis EMSURE®
Synonyms:
Hydrochloric acid solution, Hydrogen chloride solution
Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Hydrochloric acid 25%, for analysis EMSURE®