Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now
Nitrite Cell Test photometric, 1.0-90.0 mg/L (NO2-N), 3.3-295.2 mg/L (NO2-), Spectroquant®

Nitrite Test, photometric

Nitrite Cell Test photometric, 1.0-90.0 mg/L (NO2-N), 3.3-295.2 mg/L (NO2-), Spectroquant®

Products

Product Category
Brand
Product Line
Analyte Suitability
Compatibility
Detection Method
Quality Segment
Application
Reaction Type
Technique