MilliporeSigma
Phosphoric acid solution NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.

Phosphoric acid

Phosphoric acid solution NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Synonyms:
Orthophosphoric acid, Phosphoric acid
Linear Formula:
H3PO4
CAS Number:
Molecular Weight:
98.00

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique
Showing 1-20 of 23
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2