
Steritest® NEO Device

Synonym(s):
Blue Base Steritest® NEO device for sterility testing, Sterility testing device, membrane filtration device, membrane filtration canister, closed membrane filtration
Products
Product Category
Brand
Color
Quality Segment
Application
Membrane Material
Pore size (µm)
Sterility
Showing 1-19 of 19
Compare
Product Number
Description
Pricing